missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VSIV GP (strain San Juan) Recombinant Rabbit Monoclonal Antibody (110)

Rabbit Recombinant Monoclonal Antibody
159.00€ - 429.00€
Specifications
| Antigen | VSIV GP (strain San Juan) |
|---|---|
| Clone | 110 |
| Concentration | 1 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30534470
![]() |
Invitrogen™
MA560383 |
100 μL |
429.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30536210
|
Invitrogen™
MA560384 |
20 μL |
159.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VSIV GP (strain San Juan) Recombinant Monoclonal Antibody for ELISA
VSIV GP refers to the glycoprotein of the Vesicular Stomatitis Indiana Virus (VSV), a surface protein crucial for viral entry into host cells by mediating attachment and membrane fusion. It is extensively used as a vector in vaccine development, such as the rVSV-ZEBOV Ebola vaccine, and as a research tool to study viral mechanisms and immune responses.Specifications
| VSIV GP (strain San Juan) | |
| 1 mg/mL | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with no preservative | |
| Primary | |
| Protein A |
| 110 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Virus | |
| Recombinant Vesicular stomatitis Indiana virus (VSIV) (strain San Juan) glycoprotein/GP protein (Met1-Ser467). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title