Learn More
Invitrogen™ VSIV GP (strain San Juan) Recombinant Rabbit Monoclonal Antibody (110)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA560384
 View more versions of this product
Product Code. 30536210
  • 159.00€ / 20µL
Description
VSIV GP (strain San Juan) Recombinant Monoclonal Antibody for ELISA
VSIV GP refers to the glycoprotein of the Vesicular Stomatitis Indiana Virus (VSV), a surface protein crucial for viral entry into host cells by mediating attachment and membrane fusion. It is extensively used as a vector in vaccine development, such as the rVSV-ZEBOV Ebola vaccine, and as a research tool to study viral mechanisms and immune responses.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VSIV GP (strain San Juan)
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
Recombinant Vesicular stomatitis Indiana virus (VSIV) (strain San Juan) glycoprotein/GP protein (Met1-Ser467).
20 μL
Primary
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA
110
Unconjugated
Rabbit
Protein A
RUO
Virus
Antibody