Learn More
Invitrogen™ VSIV GP (strain San Juan) Recombinant Rabbit Monoclonal Antibody (110)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA560383
This item is not returnable.
View return policy
Description
VSIV GP (strain San Juan) Recombinant Monoclonal Antibody for ELISA
VSIV GP refers to the glycoprotein of the Vesicular Stomatitis Indiana Virus (VSV), a surface protein crucial for viral entry into host cells by mediating attachment and membrane fusion. It is extensively used as a vector in vaccine development, such as the rVSV-ZEBOV Ebola vaccine, and as a research tool to study viral mechanisms and immune responses.
Specifications
| VSIV GP (strain San Juan) | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Recombinant Vesicular stomatitis Indiana virus (VSIV) (strain San Juan) glycoprotein/GP protein (Met1-Ser467). | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA | |
| 110 | |
| Unconjugated | |
| Rabbit | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |