missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Seasonal H1N1 Neuraminidase Antibody (10C5F12) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
197.00€ - 509.00€
Specifications
| Antigen | Seasonal H1N1 Neuraminidase |
|---|---|
| Clone | 10C5F12 |
| Concentration | 1 mg/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18633928
|
Novus Biologicals
NBP2-41331-0.025mg |
0.025 mg |
197.00€
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18166537
|
Novus Biologicals
NBP2-41331 |
0.1 mg |
509.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Seasonal H1N1 Neuraminidase Monoclonal specifically detects Seasonal H1N1 Neuraminidase in Influenza A Virus H1N1, Virus samples. It is validated for ELISA.Specifications
| Seasonal H1N1 Neuraminidase | |
| 1 mg/mL | |
| Monoclonal | |
| Mouse | |
| Influenza virus, Virus | |
| EC=3.2.1.18, Neuraminidase | |
| IgG2b | |
| Affinity Purified |
| 10C5F12 | |
| ELISA | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| Antibody was raised against a synthetic peptide from the seasonal influenza (H1N1) neuraminidase protein. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title