missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Seasonal H1N1 Neuraminidase Antibody (10C5F12) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-41331
This item is not returnable.
View return policy
Description
Seasonal H1N1 Neuraminidase Monoclonal specifically detects Seasonal H1N1 Neuraminidase in Influenza A Virus H1N1, Virus samples. It is validated for ELISA.
Specifications
| Seasonal H1N1 Neuraminidase | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA 1 μg/mL | |
| EC=3.2.1.18, Neuraminidase | |
| Antibody was raised against a synthetic peptide from the seasonal influenza (H1N1) neuraminidase protein. | |
| 0.1 mg | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
| ELISA | |
| 10C5F12 | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Influenza virus, Virus | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction