missing translation for 'onlineSavingsMsg'
Learn More

Seasonal H1N1 Neuraminidase Antibody (10C5F12) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-41331-0.025mg

 View more versions of this product

Product Code. 18633928

  • 197.00€ / 0.03mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Seasonal H1N1 Neuraminidase Monoclonal specifically detects Seasonal H1N1 Neuraminidase in Influenza A Virus H1N1, Virus samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Seasonal H1N1 Neuraminidase
Monoclonal
1 mg/mL
ELISA 1 μg/mL
EC=3.2.1.18, Neuraminidase
Antibody was raised against a synthetic peptide from the seasonal influenza (H1N1) neuraminidase protein.
0.025 mg
Primary
Store at -20°C. Avoid freeze/thaw cycles.
ELISA
10C5F12
Unconjugated
PBS with 0.02% Sodium Azide
Mouse
Affinity Purified
RUO
Influenza virus, Virus
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.