Learn More
enQuireBio™ Recombinant Bovine LH-beta Protein
A DNA sequence encoding the Bos taurus (Bovine) LH-beta, was expressed in the hosts and tags indicated.
309.00€ - 3405.00€
Specifications
Accession Number | NP_776355.1 |
---|---|
Gene ID (Entrez) | 280839 |
Name | LH-beta Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16022320
|
enQuireBio™
QP6303-EC-100UG |
100 μg |
985.00€
100µg |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||||
16042320
|
enQuireBio™
QP6303-EC-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||||
16032320
|
enQuireBio™
QP6303-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16052320
|
enQuireBio™
QP6303-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16062320
|
enQuireBio™
QP6303-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16072320
|
enQuireBio™
QP6303-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_776355.1 | |
LH-beta Protein | |
Not determined. | |
Full Length | |
Bovine | |
His | |
Tris-based buffer, 50% glycerol |
280839 | |
Research Use Only | |
LHB | |
Recombinant Protein | |
E. coli | |
SRGPLRPLCQ PINATLAAEK EACPVCITFT TSICAGYCPS MKRVLPVILP PMPQRVCTYH ELRFASVRLP GCPPGVDPMV SFPVALSCHC GPCRLSSTDC GGPRTQPLAC DHPPLPDILF L | |
Greater than 90% as determined by SDS-PAGE. |