missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine LH-beta Protein

A DNA sequence encoding the Bos taurus (Bovine) LH-beta, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6303-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16062320

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_776355.1
LH-beta Protein
Research Use Only
LHB
Recombinant Protein
E. coli
SRGPLRPLCQ PINATLAAEK EACPVCITFT TSICAGYCPS MKRVLPVILP PMPQRVCTYH ELRFASVRLP GCPPGVDPMV SFPVALSCHC GPCRLSSTDC GGPRTQPLAC DHPPLPDILF L
Greater than 90% as determined by SDS-PAGE.
280839
500 μg
Not determined.
Full Length
Bovine
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers