Learn More
enQuireBio™ Recombinant Bovine LH-beta Protein
A DNA sequence encoding the Bos taurus (Bovine) LH-beta, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP6303-ec-200ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16052320
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
Specifications
NP_776355.1
LH-beta Protein
Research Use Only
LHB
Recombinant Protein
E. coli
SRGPLRPLCQ PINATLAAEK EACPVCITFT TSICAGYCPS MKRVLPVILP PMPQRVCTYH ELRFASVRLP GCPPGVDPMV SFPVALSCHC GPCRLSSTDC GGPRTQPLAC DHPPLPDILF L
Greater than 90% as determined by SDS-PAGE.
280839
200 μg
Not determined.
Full Length
Bovine
His
Tris-based buffer, 50% glycerol