missing translation for 'onlineSavingsMsg'
Learn More

PXR/NR1I2 Antibody (6H11D8) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

203.00€ - 552.00€

Specifications

Antigen PXR/NR1I2
Clone 6H11D8
Concentration 1 mg/mL
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000
Applications Western Blot, Flow Cytometry, ELISA
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18264252
View Documents
Novus Biologicals
NBP2-61817
0.1 mL
552.00€
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18699006
View Documents
Novus Biologicals
NBP2-61817-0.025ml
0.025 mL
203.00€
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PXR/NR1I2 Monoclonal specifically detects PXR/NR1I2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications

Specifications

PXR/NR1I2
1 mg/mL
Western Blot, Flow Cytometry, ELISA
Unconjugated
Mouse
nuclear receptor subfamily 1 group I member 2, nuclear receptor subfamily 1, group I, member 2, ONR1, Orphan nuclear receptor PAR1, Orphan nuclear receptor PXR, PAR, PAR1, PAR2, Pregnane X receptor, PRR, PXRPARq, SAR, Steroid and xenobiotic receptor, SXRBXR
NR1I2
IgG1
Protein G purified
6H11D8
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000
Monoclonal
Purified
Stem Cell Markers, Transcription Factors and Regulators
8856
Purified recombinant fragment of human NR1I2 (AA:1-142) expressed in E. Coli.
Primary
9.8 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.