missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PXR/NR1I2 Monoclonal specifically detects PXR/NR1I2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
Specifications
| Antigen | PXR/NR1I2 |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Clone | 6H11D8 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | nuclear receptor subfamily 1 group I member 2, nuclear receptor subfamily 1, group I, member 2, ONR1, Orphan nuclear receptor PAR1, Orphan nuclear receptor PXR, PAR, PAR1, PAR2, Pregnane X receptor, PRR, PXRPARq, SAR, Steroid and xenobiotic receptor, SXRBXR |
| Gene Symbols | NR1I2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?