missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PXR/NR1I2 Antibody (6H11D8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61817-0.025ml
This item is not returnable.
View return policy
Description
PXR/NR1I2 Monoclonal specifically detects PXR/NR1I2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| PXR/NR1I2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 | |
| nuclear receptor subfamily 1 group I member 2, nuclear receptor subfamily 1, group I, member 2, ONR1, Orphan nuclear receptor PAR1, Orphan nuclear receptor PXR, PAR, PAR1, PAR2, Pregnane X receptor, PRR, PXRPARq, SAR, Steroid and xenobiotic receptor, SXRBXR | |
| Mouse | |
| 9.8 kDa | |
| 0.025 mL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA | |
| 6H11D8 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| NR1I2 | |
| Purified recombinant fragment of human NR1I2 (AA:1-142) expressed in E. Coli. | |
| Protein G purified | |
| Stem Cell Markers, Transcription Factors and Regulators | |
| 8856 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction