Learn More
Porcelaines Avignon™ Porcelain Buchner Funnel
The Porcelain Buchner Funnel is chemically neutral and has a high resistance to thermal and physical shocks.
19.78€ - 25.80€
Product Code | Brand | Diameter (Metric) Plate | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Plate | Price | Quantity & Availability | |||||
11788403
|
Porcelaines Avignon™
12702 |
30mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
11708403
|
Porcelaines Avignon™
12703 |
40mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
11768403
|
Porcelaines Avignon™
12704 |
50mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Description
- High resistance to thermal and physical shock
- Can be used up to 1100°C
- Chemically neutral
Specifications
Porcelain | |
1Each | |
Buchner Funnel |