missing translation for 'onlineSavingsMsg'
Learn More

Porcelaines Avignon™ Porcelain Buchner Funnel

The Porcelain Buchner Funnel is chemically neutral and has a high resistance to thermal and physical shocks.

Brand:  Porcelaines Avignon™ 12703

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11708403

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

  • High resistance to thermal and physical shock
  • Can be used up to 1100°C
  • Chemically neutral
Specifications

Specifications

Buchner Funnel
82 mm
40mm
1Each
Porcelain
40mm
82mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers