Learn More
Porcelaines Avignon™ Porcelain Buchner Funnel
The Porcelain Buchner Funnel is chemically neutral and has a high resistance to thermal and physical shocks.
Brand: Porcelaines Avignon™ 12702
Additional Details : Weight : 0.01800kg
Description
- High resistance to thermal and physical shock
- Can be used up to 1100°C
- Chemically neutral
Specifications
Buchner Funnel | |
59 mm | |
30mm | |
1Each |
Porcelain | |
30mm | |
59mm |