missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Porcelaines Avignon™ Porcelain Buchner Funnel
The Porcelain Buchner Funnel is chemically neutral and has a high resistance to thermal and physical shocks.
Brand: Porcelaines Avignon™ 12704
Additional Details : Weight : 0.80000kg
Description
- High resistance to thermal and physical shock
- Can be used up to 1100°C
- Chemically neutral
Specifications
Buchner Funnel | |
95 mm | |
50mm |
Porcelain | |
50mm | |
1Each |