missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OCT4 Antibody (7E7), Novus Biologicals™
Mouse Monoclonal Antibody
231.00€ - 484.00€
Specifications
| Antigen | OCT4 |
|---|---|
| Clone | 70000000 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18247192
|
Novus Biologicals
NBP1-51664SS |
0.025 mL |
231.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18273412
|
Novus Biologicals
NBP1-51664 |
0.1 mL |
484.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
44473 Monoclonal specifically detects 44473 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| OCT4 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| Q01860 | |
| 5460 | |
| Synthetic peptide derived from an internal portion of human OCT4. [Uniprot: Q01860] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 70000000 | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cellular Markers, Chromatin Research, Embryonic Stem Cell Markers, Glia Markers, Neuroscience, Oligodendrocyte Cell Markers, Pancreatic Stem Cell Markers, Stem Cell Markers, Transcription Factors and Regulators | |
| Ascites with 0.03% Sodium Azide | |
| class 5, transcription factor 1, MGC22487, Oct-3, OCT3Oct4, Oct-4, Octamer-binding protein 3, otc-4, OTF3POU domain class 5, transcription factor 1, OTF4, POU class 5 homeobox 1, POU-type homeodomain-containing DNA-binding protein | |
| POU5F1 | |
| IgG1 | |
| Unpurified | |
| 45 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title