missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OCT4 Antibody (7E7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-51664SS
This item is not returnable.
View return policy
Description
44473 Monoclonal specifically detects 44473 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| OCT4 | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.03% Sodium Azide | |
| class 5, transcription factor 1, MGC22487, Oct-3, OCT3Oct4, Oct-4, Octamer-binding protein 3, otc-4, OTF3POU domain class 5, transcription factor 1, OTF4, POU class 5 homeobox 1, POU-type homeodomain-containing DNA-binding protein | |
| Mouse | |
| 45 kDa | |
| 0.025 mL | |
| Cellular Markers, Chromatin Research, Embryonic Stem Cell Markers, Glia Markers, Neuroscience, Oligodendrocyte Cell Markers, Pancreatic Stem Cell Markers, Stem Cell Markers, Transcription Factors and Regulators | |
| 5460 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 70000000 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 | |
| Q01860 | |
| POU5F1 | |
| Synthetic peptide derived from an internal portion of human OCT4. [Uniprot: Q01860] | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction