missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INSIG-1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
219.45€ - 509.25€
Specifications
| Antigen | INSIG-1 |
|---|---|
| Concentration | 1.02 mg/mL |
| Dilution | Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 1:100-1:1000 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18036564
|
Novus Biologicals
NB110-55244SS |
0.025 mL |
232.00€ 219.45€ / 25µL Save 12.55€ 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18737183
|
Novus Biologicals
NB110-55244 |
0.1 mL |
539.00€ 509.25€ / 0.10mL Save 29.75€ 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
INSIG-1 Polyclonal specifically detects INSIG-1 in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| INSIG-1 | |
| Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
| Polyclonal | |
| Rabbit | |
| Cholesterol Metabolism, ER Markers, Lipid and Metabolism, Signal Transduction | |
| PBS and 30% Glycerol with 0.1% Sodium Azide | |
| CL6, CL-6, INSIG-1, INSIG-1 membrane protein, insulin induced gene 1, insulin-induced gene 1 protein, MGC1405 | |
| INSIG1 | |
| IgG | |
| Affinity Purified | |
| This is specific for Insig1 but not Insig2. |
| 1.02 mg/mL | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human, Rat | |
| O15503 | |
| 3638 | |
| A synthetic peptide representing an internal region of the human INSIG-1 protein (between residues 1-100) according to Swiss-Prot# O15503. | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| 30 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title