missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INSIG-1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NB110-55244
This item is not returnable.
View return policy
Description
INSIG-1 Polyclonal specifically detects INSIG-1 in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| INSIG-1 | |
| Polyclonal | |
| Unconjugated | |
| PBS and 30% Glycerol with 0.1% Sodium Azide | |
| CL6, CL-6, INSIG-1, INSIG-1 membrane protein, insulin induced gene 1, insulin-induced gene 1 protein, MGC1405 | |
| Rabbit | |
| 30 kDa | |
| 0.1 mL | |
| Cholesterol Metabolism, ER Markers, Lipid and Metabolism, Signal Transduction | |
| 3638 | |
| Human, Rat | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 1.02 mg/mL | |
| Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
| O15503 | |
| INSIG1 | |
| A synthetic peptide representing an internal region of the human INSIG-1 protein (between residues 1-100) according to Swiss-Prot# O15503. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This is specific for Insig1 but not Insig2. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction