missing translation for 'onlineSavingsMsg'
Learn More

INSIG-1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications

Brand:  Novus Biologicals NB110-55244SS

 View more versions of this product

Product Code. 18036564

  • ONLINE EXCLUSIVE PRICE
    232.00€
    219.45€ / 25µL
    Save 12.55€
    5% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

INSIG-1 Polyclonal specifically detects INSIG-1 in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

INSIG-1
Polyclonal
Unconjugated
PBS and 30% Glycerol with 0.1% Sodium Azide
CL6, CL-6, INSIG-1, INSIG-1 membrane protein, insulin induced gene 1, insulin-induced gene 1 protein, MGC1405
Rabbit
30 kDa
0.025 mL
Cholesterol Metabolism, ER Markers, Lipid and Metabolism, Signal Transduction
3638
Human, Rat
IgG
Western Blot, Immunocytochemistry, Immunofluorescence
1.02 mg/mL
Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 1:100-1:1000
O15503
INSIG1
A synthetic peptide representing an internal region of the human INSIG-1 protein (between residues 1-100) according to Swiss-Prot# O15503.
Affinity Purified
RUO
Primary
This is specific for Insig1 but not Insig2.
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only