missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Kininogen Antibody, R&D Systems™

Goat Polyclonal Antibody
203.00€ - 501.00€
Specifications
| Antigen | Kininogen |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18708137
![]() |
R&D Systems
AF1569-SP |
25 μg |
203.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18013022
|
R&D Systems
AF1569 |
100 μg |
501.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Kininogen Polyclonal specifically detects Kininogen in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Kininogen | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| 3827 | |
| Mouse myeloma cell line NS0-derived recombinant human Kininogen Gln19-Ser644 Accession # P01042 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human Kininogen in direct ELISAs and Western blots. |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| P01042 | |
| KNG1 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title