missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Kininogen Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1569-SP
This item is not returnable.
View return policy
Description
Kininogen Polyclonal specifically detects Kininogen in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Kininogen | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| KNG1 | |
| Mouse myeloma cell line NS0-derived recombinant human Kininogen Gln19-Ser644 Accession # P01042 | |
| 25 μg | |
| Primary | |
| Detects human Kininogen in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| P01042 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3827 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction