missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Kininogen Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF1569
396.95 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
Kininogen Polyclonal specifically detects Kininogen in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Kininogen | |
| Polyclonal | |
| Unconjugated | |
| P01042 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3827 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| KNG1 | |
| Mouse myeloma cell line NS0-derived recombinant human Kininogen Gln19-Ser644 Accession # P01042 | |
| 100 μg | |
| Primary | |
| Detects human Kininogen in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction