missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human EAAT1/GLAST-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
209.00€ - 444.00€
Specifications
| Antigen | EAAT1/GLAST-1/SLC1A3 |
|---|---|
| Clone | 482420 |
| Dilution | Immunohistochemistry 5-25 ug/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18771957
![]() |
R&D Systems
MAB6048-SP |
25 μg |
209.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18713907
![]() |
R&D Systems
MAB6048-100 |
100 μg |
444.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EAAT1/GLAST-1/SLC1A3 Monoclonal specifically detects EAAT1/GLAST-1/SLC1A3 in Human samples. It is validated for Immunohistochemistry.Specifications
| EAAT1/GLAST-1/SLC1A3 | |
| Immunohistochemistry 5-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| EA6, EA6FLJ25094, EAAT1GLAST-1, excitatory amino acid transporter 1, GLAST1, GLAST-1, GLASTGLAST1, SLC1A3, Sodium-dependent glutamate/aspartate transporter 1, solute carrier family 1 (glial high affinity glutamate transporter), member 3, Solute carrier family 1 member 3 | |
| E. coli-derived recombinant human EAAT1/GLAST-1, His146-Ser237, Accession # P43003 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human EAAT1/GLAST-1 in direct ELISAs. |
| 482420 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P43003 | |
| 6507 | |
| IgG2b | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title