missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human EAAT1/GLAST-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6048-100
This item is not returnable.
View return policy
Description
EAAT1/GLAST-1/SLC1A3 Monoclonal specifically detects EAAT1/GLAST-1/SLC1A3 in Human samples. It is validated for Immunohistochemistry.
Specifications
| EAAT1/GLAST-1/SLC1A3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| EA6, EA6FLJ25094, EAAT1GLAST-1, excitatory amino acid transporter 1, GLAST1, GLAST-1, GLASTGLAST1, SLC1A3, Sodium-dependent glutamate/aspartate transporter 1, solute carrier family 1 (glial high affinity glutamate transporter), member 3, Solute carrier family 1 member 3 | |
| E. coli-derived recombinant human EAAT1/GLAST-1, His146-Ser237, Accession # P43003 | |
| 100 μg | |
| Primary | |
| Detects human EAAT1/GLAST-1 in direct ELISAs. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 482420 | |
| Immunohistochemistry 5-25 ug/mL | |
| P43003 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 6507 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction