missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Desmin Antibody, Novus Biologicals Biologicals™
Rabbit Monoclonal Antibody
247.00€ - 572.00€
Specifications
| Antigen | Desmin |
|---|---|
| Clone | 3112A |
| Dilution | Western Blot 1 μg/mL, Immunohistochemistry 1-10 μg/mL, Multiplex Immunofluorescence 0.1 μg/mL |
| Applications | Multiplex Immunofluorescence, Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30484811
|
R&D Systems
MAB11654-100 |
100 μg |
572.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30484812
|
R&D Systems
MAB11654-SP |
25 μg |
247.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Desmin Monoclonal antibody specifically detects Desmin in Human samples. It is validated for Multiplex Immunofluorescence,Western Blot,ImmunohistochemistrySpecifications
| Desmin | |
| Western Blot 1 μg/mL, Immunohistochemistry 1-10 μg/mL, Multiplex Immunofluorescence 0.1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. | |
| CMD1I, CMD1IFLJ41013, CSM1, CSM2, DES, desmin, FLJ12025, FLJ39719, FLJ41793, intermediate filament protein | |
| E. coli- derived recombinant human Desmin, Val261-Leu470, Accession # P17661 | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. | |
| Antibody |
| 3112A | |
| Multiplex Immunofluorescence, Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P17661 | |
| 1674 | |
| IgG | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title