Learn More
Human Desmin Antibody, Novus Biologicals Biologicals™
Rabbit Monoclonal Antibody
Brand:  R&D Systems MAB11654-SP
 View more versions of this product
Product Code. 30484812
  • 247.00€ / 25µg
Description
Desmin Monoclonal antibody specifically detects Desmin in Human samples. It is validated for Multiplex Immunofluorescence,Western Blot,Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Desmin
Monoclonal
Unconjugated
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
CMD1I, CMD1IFLJ41013, CSM1, CSM2, DES, desmin, FLJ12025, FLJ39719, FLJ41793, intermediate filament protein
E. coli- derived recombinant human Desmin, Val261-Leu470, Accession # P17661
25 μg
Primary
Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Purified
Multiplex Immunofluorescence, Western Blot, Immunohistochemistry
3112A
Western Blot 1 μg/mL, Immunohistochemistry 1-10 μg/mL, Multiplex Immunofluorescence 0.1 μg/mL
P17661
Rabbit
Protein A or G purified from hybridoma culture supernatant
RUO
1674
Human
Antibody