Learn More
Human Desmin Antibody, Novus Biologicals Biologicals™
Rabbit Monoclonal Antibody
Brand: R&D Systems MAB11654-100
This item is not returnable.
View return policy
Description
Desmin Monoclonal antibody specifically detects Desmin in Human samples. It is validated for Multiplex Immunofluorescence,Western Blot,Immunohistochemistry
Specifications
| Desmin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. | |
| CMD1I, CMD1IFLJ41013, CSM1, CSM2, DES, desmin, FLJ12025, FLJ39719, FLJ41793, intermediate filament protein | |
| E. coli- derived recombinant human Desmin, Val261-Leu470, Accession # P17661 | |
| 100 μg | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| Purified |
| Multiplex Immunofluorescence, Western Blot, Immunohistochemistry | |
| 3112A | |
| Western Blot 1 μg/mL, Immunohistochemistry 1-10 μg/mL, Multiplex Immunofluorescence 0.1 μg/mL | |
| P17661 | |
| Rabbit | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1674 | |
| Human | |
| Antibody | |
| IgG |