missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Doublecortin Antibody (3E1), Novus Biologicals™
Mouse Monoclonal Antibody
209.00€ - 511.00€
Specifications
| Applications | Western Blot |
|---|---|
| Classification | Monoclonal |
| Target Species | Human, Pig |
| Content And Storage | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Product Code | Brand | dimQuantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | dimQuantity | Price | Quantity & Availability | |||||
|
18700534
|
Novus Biologicals
NBP1-92684SS |
209.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18780294
|
Novus Biologicals
NBP1-92684 |
511.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Doublecortin Monoclonal specifically detects Doublecortin in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| Western Blot | |
| Monoclonal | |
| Human, Pig | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title