Learn More
Doublecortin Antibody (3E1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP1-92684
 View more versions of this product
Product Code. 18780294
  • 511.00€ / 0.10mL
Description
Doublecortin Monoclonal specifically detects Doublecortin in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Monoclonal
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG2a
Mouse, Rat, Pig