Learn More
Doublecortin Antibody (3E1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP1-92684SS
 View more versions of this product
Product Code. 18700534
  • 209.00€ / 25µL
Description
Doublecortin Monoclonal specifically detects Doublecortin in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Western Blot
Monoclonal
Human, Pig
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.