Learn More
Doublecortin Antibody (3E1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-92684SS
This item is not returnable.
View return policy
Description
Doublecortin Monoclonal specifically detects Doublecortin in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Western Blot | |
| Monoclonal | |
| Human, Pig | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |