missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 10 Antibody (1C3D9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
205.00€ - 552.00€
Specifications
| Antigen | Cytokeratin 10 |
|---|---|
| Clone | 1C3D9 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:200-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18285511
|
Novus Biologicals
NBP2-61737 |
0.1 mL |
552.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18699957
|
Novus Biologicals
NBP2-61737-0.025ml |
0.025 mL |
205.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytokeratin 10 Monoclonal specifically detects Cytokeratin 10 in Human, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Cytokeratin 10 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human, Monkey | |
| 3858 | |
| Purified recombinant fragment of human KRT10 (AA: 345-454 ) expressed in E. Coli. | |
| Primary | |
| 176 kDa |
| 1C3D9 | |
| Western Blot 1:200-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Monoclonal | |
| Purified | |
| Cytoskeleton Markers | |
| BCIE, BIE, CK10, CK-10, cytokeratin 10, Cytokeratin-10, EHK, K10keratosis palmaris et plantaris, keratin 10, Keratin 10 (epidermolytic hyperkeratosis; keratosis palmaris et plantaris), keratin, type I cytoskeletal 10, keratin-10, KPP | |
| KRT10 | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title