missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 10 Antibody (1C3D9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-61737

 View more versions of this product

Product Code. 18285511

  • 552.00€ / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Cytokeratin 10 Monoclonal specifically detects Cytokeratin 10 in Human, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cytokeratin 10
Monoclonal
1 mg/mL
Western Blot 1:200-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000
BCIE, BIE, CK10, CK-10, cytokeratin 10, Cytokeratin-10, EHK, K10keratosis palmaris et plantaris, keratin 10, Keratin 10 (epidermolytic hyperkeratosis; keratosis palmaris et plantaris), keratin, type I cytoskeletal 10, keratin-10, KPP
Mouse
176 kDa
0.1 mL
Primary
Human, Monkey
Purified
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
1C3D9
Unconjugated
PBS with 0.05% Sodium Azide
KRT10
Purified recombinant fragment of human KRT10 (AA: 345-454 ) expressed in E. Coli.
Protein G purified
Cytoskeleton Markers
3858
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.