missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 10 Antibody (1C3D9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-61737-0.025ml

 View more versions of this product

Product Code. 18699957

  • 205.00€ / 0.03mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Cytokeratin 10 Monoclonal specifically detects Cytokeratin 10 in Human, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cytokeratin 10
Monoclonal
1 mg/mL
Western Blot 1:200-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000
BCIE, BIE, CK10, CK-10, cytokeratin 10, Cytokeratin-10, EHK, K10keratosis palmaris et plantaris, keratin 10, Keratin 10 (epidermolytic hyperkeratosis; keratosis palmaris et plantaris), keratin, type I cytoskeletal 10, keratin-10, KPP
Mouse
176 kDa
0.025 mL
Primary
Human, Monkey
Purified
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
1C3D9
Unconjugated
PBS with 0.05% Sodium Azide
KRT10
Purified recombinant fragment of human KRT10 (AA: 345-454 ) expressed in E. Coli.
Protein G purified
Cytoskeleton Markers
3858
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.