Learn More
cis-3-Hexenyl acetate, 98+%
Product Code. 11443977
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11443977 100 g 100g
11433977 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11443977 Supplier Thermo Scientific Alfa Aesar Supplier No. A19380.22
Request Bulk or Custom Format
CAS: 3681-71-8 | C8H14O2 | 142.198 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3681-71-8
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.198
MDL Number MFCD00036563
InChI Key NPFVOOAXDOBMCE-PLNGDYQASA-N
Synonym cis-3-hexenyl acetate, z-3-hexenyl acetate, z-hex-3-enyl acetate, 3-hexenylacetate, cis-3-hexenyl ethanoate, cis-3-hexen-1-yl acetate, leaf acetate, 3-hexen-1-ol, acetate, z, cis-3-hexenol acetate, 3z-hex-3-en-1-yl acetate
PubChem CID 5363388
ChEBI CHEBI:61316
IUPAC Name [(Z)-hex-3-enyl] acetate
SMILES CCC=CCCOC(=O)C
Specifications
Density 0.899
Boiling Point 169°C
Flash Point 59°C (138°F)
Refractive Index 1.427
Quantity 100 g
UN Number UN3272
Beilstein 1721854
Formula Weight 142.2
Percent Purity 99%
Chemical Name or Material cis-3-Hexenyl acetate