CAS RN 3681-71-8
CAS RN 3681-71-8
IUPAC Name:
(3Z)-hex-3-en-1-yl acetate
Synonyms:
(Z)-3-hexenyl acetate
Molecular Weight (g/mol):
142.2
Molecular Formula:
C8H14O2
InChi Key:
NPFVOOAXDOBMCE-PLNGDYQASA-N
SMILES:
CC\C=C/CCOC(C)=O
1
–
1
of
1
results
1
cis-3-Hexenyl acetate, 98+%
CAS: 3681-71-8 Molecular Formula: C8H14O2 Molecular Weight (g/mol): 142.198 MDL Number: MFCD00036563 InChI Key: NPFVOOAXDOBMCE-PLNGDYQASA-N Synonym: cis-3-hexenyl acetate,z-3-hexenyl acetate,z-hex-3-enyl acetate,3-hexenylacetate,cis-3-hexenyl ethanoate,cis-3-hexen-1-yl acetate,leaf acetate,3-hexen-1-ol, acetate, z,cis-3-hexenol acetate,3z-hex-3-en-1-yl acetate PubChem CID: 5363388 ChEBI: CHEBI:61316 IUPAC Name: [(Z)-hex-3-enyl] acetate SMILES: CCC=CCCOC(=O)C