missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Rel Antibody (1E7), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
230.00€ - 578.00€
Specifications
| Antigen | c-Rel |
|---|---|
| Clone | 10000000 |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18161178
|
Novus Biologicals
NBP2-37593 |
0.1 mL |
578.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18660366
|
Novus Biologicals
NBP2-37593-0.025ml |
0.025 mL |
230.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
c-Rel Monoclonal specifically detects c-Rel in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| c-Rel | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human, Mouse | |
| C-Rel, C-Rel proto-oncogene protein, I-Rel, oncogene REL, avian reticuloendotheliosis, proto-oncogene c-Rel, v-rel avian reticuloendotheliosis viral oncogene homolog, v-rel reticuloendotheliosis viral oncogene homolog (avian) | |
| REL | |
| IgG1 | |
| Unpurified |
| 10000000 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors | |
| Ascites with 0.03% Sodium Azide | |
| 5966 | |
| Purified recombinant fragment of human c-Rel expressed in E. Coli. | |
| Primary | |
| 68.5 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title