Learn More
c-Rel Antibody (1E7), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand:  Novus Biologicals NBP2-37593
 View more versions of this product
Product Code. 18161178
  • 578.00€ / 0.10mL
Description
c-Rel Monoclonal specifically detects c-Rel in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
c-Rel
Monoclonal
Unconjugated
Ascites with 0.03% Sodium Azide
REL
Purified recombinant fragment of human c-Rel expressed in E. Coli.
Unpurified
RUO
Primary
Human, Mouse
Ascites
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
10000000
Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000
C-Rel, C-Rel proto-oncogene protein, I-Rel, oncogene REL, avian reticuloendotheliosis, proto-oncogene c-Rel, v-rel avian reticuloendotheliosis viral oncogene homolog, v-rel reticuloendotheliosis viral oncogene homolog (avian)
Mouse
68.5 kDa
0.1 mL
Cancer, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors
5966
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.