missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Rel Antibody (1E7), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-37593-0.025ml
This item is not returnable.
View return policy
Description
c-Rel Monoclonal specifically detects c-Rel in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| c-Rel | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.03% Sodium Azide | |
| REL | |
| Purified recombinant fragment of human c-Rel expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Ascites |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 10000000 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| C-Rel, C-Rel proto-oncogene protein, I-Rel, oncogene REL, avian reticuloendotheliosis, proto-oncogene c-Rel, v-rel avian reticuloendotheliosis viral oncogene homolog, v-rel reticuloendotheliosis viral oncogene homolog (avian) | |
| Mouse | |
| 68.5 kDa | |
| 0.025 mL | |
| Cancer, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors | |
| 5966 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction