missing translation for 'onlineSavingsMsg'
Learn More

APP Antibody (S05-3E3), Novus Biologicals™

Rabbit Monoclonal Antibody

421.00€ - 645.00€

Specifications

Antigen APP
Clone S05-3E3
Concentration 0.3 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18635574
View Documents
Novus Biologicals
NBP3-20015-50ul
50 μg
421.00€
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18644115
View Documents
Novus Biologicals
NBP3-20015-100ul
100 μg
645.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

APP Monoclonal antibody specifically detects APP in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

APP
0.3 mg/mL
Monoclonal
Purified
RUO
Human, Mouse, Rat
amyloid beta (A4) precursor protein-binding, family B, member 2, amyloid beta A4 precursor protein-binding family B member 2, Amyloid beta precursor protein, APP
A synthetic peptide of human APP (Uniprot # P05067)
Primary
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
S05-3E3
Western Blot
Unconjugated
Rabbit
Alzheimers Research, Apoptosis, Cell Cycle and Replication, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
351
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.