missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APP Antibody (S05-3E3), Novus Biologicals™
Rabbit Monoclonal Antibody
Marca: Novus Biologicals NBP3-20015-100ul
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
APP Monoclonal antibody specifically detects APP in Human, Mouse, Rat samples. It is validated for Western Blot
Specifica
| APP | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S05-3E3 | |
| Unconjugated | |
| amyloid beta (A4) precursor protein-binding, family B, member 2, amyloid beta A4 precursor protein-binding family B member 2, Amyloid beta precursor protein, APP | |
| A synthetic peptide of human APP (Uniprot # P05067) | |
| 100 μg | |
| Alzheimers Research, Apoptosis, Cell Cycle and Replication, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific | |
| 351 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto