Learn More
APP Antibody (S05-3E3), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-20015-50ul
 View more versions of this product
Product Code. 18635574
  • 421.00€ / 50µL
Description
APP Monoclonal antibody specifically detects APP in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
APP
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot
S05-3E3
Unconjugated
amyloid beta (A4) precursor protein-binding, family B, member 2, amyloid beta A4 precursor protein-binding family B member 2, Amyloid beta precursor protein, APP
A synthetic peptide of human APP (Uniprot # P05067)
50 μg
Alzheimers Research, Apoptosis, Cell Cycle and Replication, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific
351
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.