missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta Amyloid Antibody (MOAB-2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 34 publications
268.00€ - 580.00€
Specifications
| Applications | Immunofluorescence, Immunoprecipitation |
|---|---|
| Classification | Monoclonal |
| Form | Purified |
| Target Species | Human, Rat, Bacteria, Monkey |
| Immunogen | This beta Amyloid antibody was developed against recombinant human beta Amyloid 42. |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18787523
|
Novus Biologicals
NBP2-13075SS |
0.025 mL |
268.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18755383
|
Novus Biologicals
NBP2-13075 |
580.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
beta Amyloid Monoclonal specifically detects beta Amyloid in Human, Mouse, Rat, Bacteria, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Dot Blot, Immunohistochemistry Free-Floating.Specifications
| Immunofluorescence, Immunoprecipitation | |
| Purified | |
| This beta Amyloid antibody was developed against recombinant human beta Amyloid 42. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Monoclonal | |
| Human, Rat, Bacteria, Monkey | |
| IgG2b |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title