Learn More
beta Amyloid Antibody (MOAB-2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 34 publications
Brand:  Novus Biologicals NBP2-13075SS
 View more versions of this product
Product Code. 18787523
  • 268.00€ / 25µL
Description
beta Amyloid Monoclonal specifically detects beta Amyloid in Human, Mouse, Rat, Bacteria, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Dot Blot, Immunohistochemistry Free-Floating.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Immunofluorescence, Immunoprecipitation
This beta Amyloid antibody was developed against recombinant human beta Amyloid 42.
Human, Rat, Bacteria, Monkey
Purified
Monoclonal
0.025 mL
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG2b