Learn More
beta Amyloid Antibody (MOAB-2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 34 publications
Brand:  Novus Biologicals NBP2-13075
 View more versions of this product
Product Code. 18755383
  • 580.00€ / 0.10mL
Description
beta Amyloid Monoclonal specifically detects beta Amyloid in Human, Mouse, Rat, Bacteria, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Dot Blot, Immunohistochemistry Free-Floating.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Monoclonal
Purified
IgG2b