missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGR3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | AGR3 |
|---|---|
| Concentration | 0.2 mg/mL |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16839393
|
Proteintech
11967-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16829393
|
Proteintech
11967-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AGR3 (Anterior Gradient 3) protein, also known as AG3 (hAG3, HAG3 in human), or BCMP11, is a secreted cytoplasmic protein which is involved in metastasis induction and p53 tumor suppressor inhibition. It may serve as molecular marker and potential therapeutic target for hormone-responsive breast tumors. Its Xenopus homolog is associated with anteroposterior fate determination during early development.Specifications
| AGR3 | |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Q8R3W7, Q8TD06 | |
| 155465, 403205 | |
| AGR3 Fusion Protein Ag2577 | |
| Primary | |
| -20°C | |
| AGR3 |
| 0.2 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| AG 3, AG3, AGR3, hAG 3, HAG3, PDIA18 | |
| Agr3 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title