missing translation for 'onlineSavingsMsg'
Learn More

AGR3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Product Code. 16839393 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16839393 20 μL 20µL
16829393 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16839393 Supplier Proteintech Supplier No. 119671AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AGR3 (Anterior Gradient 3) protein, also known as AG3 (hAG3, HAG3 in human), or BCMP11, is a secreted cytoplasmic protein which is involved in metastasis induction and p53 tumor suppressor inhibition. It may serve as molecular marker and potential therapeutic target for hormone-responsive breast tumors. Its Xenopus homolog is associated with anteroposterior fate determination during early development.

Specifications

Antigen AGR3
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene AGR3
Gene Accession No. Q8R3W7, Q8TD06
Gene Alias AG 3, AG3, AGR3, hAG 3, HAG3, PDIA18
Gene Symbols Agr3
Host Species Rabbit
Immunogen AGR3 Fusion Protein Ag2577
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 155465, 403205
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.