missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
AGR3 (Anterior Gradient 3) protein, also known as AG3 (hAG3, HAG3 in human), or BCMP11, is a secreted cytoplasmic protein which is involved in metastasis induction and p53 tumor suppressor inhibition. It may serve as molecular marker and potential therapeutic target for hormone-responsive breast tumors. Its Xenopus homolog is associated with anteroposterior fate determination during early development.
Specifications
Specifications
| Antigen | AGR3 |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | AGR3 |
| Gene Accession No. | Q8R3W7, Q8TD06 |
| Gene Alias | AG 3, AG3, AGR3, hAG 3, HAG3, PDIA18 |
| Gene Symbols | Agr3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?