missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCG8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 7 publications
213.00€ - 493.00€
Specifications
| Antigen | ABCG8 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:250-1:500, Immunocytochemistry/Immunofluorescence 1:250 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18290001
|
Novus Biologicals
NB400-117SS |
0.05 mL |
213.00€
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18711684
|
Novus Biologicals
NB400-117 |
493.00€
0.20mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
ABCG8 Polyclonal specifically detects ABCG8 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| ABCG8 | |
| Western Blot 1:250-1:500, Immunocytochemistry/Immunofluorescence 1:250 | |
| Polyclonal | |
| Rabbit | |
| ABC Transporters, Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.02% Sodium Azide | |
| ATP-binding cassette sub-family G member 8, ATP-binding cassette, sub-family G (WHITE), member 8, ATP-binding cassette, sub-family G (WHITE), member 8 (sterolin 2), GBD4ATP-binding cassette, subfamily G, member 8, MGC142217, sterolin 2, sterolin-2, STSL | |
| ABCG8 | |
| IgG | |
| Affinity Purified | |
| 70 kDa |
| 1 mg/mL | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| Q9H221 | |
| 64241 | |
| A synthetic peptide from the N-terminal region of human ABCG8 protein. [UniProt# Q9H221] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title